Product Details

SNP ID
rs140050111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99619985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCTGACAGAAAGAGCACTGGAG[A/G]CCAAGGCGGTGGGTGAGGAGGGGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GS1-259H13.2 PubMed Links

Gene Details

Gene
GS1-259H13.2
Gene Name
transmembrane protein 225-like
There are no transcripts associated with this gene.

Gene
ZSCAN25
Gene Name
zinc finger and SCAN domain containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145115.2 961 Missense Mutation ACC,GCC T127A NP_660090.2
XM_005250194.2 961 Missense Mutation ACC,GCC T127A XP_005250251.1
XM_011515905.2 961 Missense Mutation ACC,GCC T127A XP_011514207.1
XM_011515906.2 961 Missense Mutation ACC,GCC T127A XP_011514208.1
XM_011515907.2 961 Missense Mutation ACC,GCC T127A XP_011514209.1
XM_011515908.2 961 UTR 5 XP_011514210.1
XM_011515909.2 961 Missense Mutation ACC,GCC T127A XP_011514211.1
XM_011515910.2 961 Missense Mutation ACC,GCC T127A XP_011514212.1
XM_017011824.1 961 Missense Mutation ACC,GCC T127A XP_016867313.1
XM_017011825.1 961 UTR 5 XP_016867314.1
XM_017011826.1 961 UTR 5 XP_016867315.1
XM_017011827.1 961 Missense Mutation ACC,GCC T127A XP_016867316.1

View Full Product Details