Product Details

SNP ID
rs140464086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101316463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATACCACATCTCCATTTCCTTCC[G/T]GTGGCTTGGGATGTCAGCATTGAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IFT22 PubMed Links

Gene Details

Gene
IFT22
Gene Name
intraflagellar transport 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130820.2 473 Silent Mutation GGG,TGG G66W NP_001124292.1
NM_001130821.2 473 Silent Mutation GGG,TGG G19W NP_001124293.1
NM_001130822.2 473 Silent Mutation GGG,TGG G19W NP_001124294.1
NM_001287525.1 473 Silent Mutation GGG,TGG G19W NP_001274454.1
NM_001287526.1 473 Silent Mutation GGG,TGG G19W NP_001274455.1
NM_022777.3 473 Silent Mutation GGG,TGG G96W NP_073614.1
Gene
LOC101927746
Gene Name
uncharacterized LOC101927746
There are no transcripts associated with this gene.

View Full Product Details