Product Details

SNP ID
rs141050594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:80136402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTTTCAGTAAGCTTCTGTTTCA[C/T]TGGTTTAATGACTGCAGAAAAAATG
Phenotype
MIM: 139310
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNAI1 PubMed Links

Gene Details

Gene
GNAI1
Gene Name
G protein subunit alpha i1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256414.1 Intron NP_001243343.1
NM_002069.5 Intron NP_002060.4

View Full Product Details