Product Details

SNP ID
rs143984888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122995046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTCAGTGATGCCATGAGAAAGA[C/T]GGTGGAGGCCAGGAACAGGATGAAA
Phenotype
MIM: 604867
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAS2R16 PubMed Links

Gene Details

Gene
TAS2R16
Gene Name
taste 2 receptor member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016945.2 655 Missense Mutation ATC,GTC I197V NP_058641.1

View Full Product Details