Product Details
- SNP ID
-
rs144989302
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:44796739 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTATTAGCCATGGTCAACCCCAC[C/T]GTGTTCTTCGACATTGCCGTCGACG
- Phenotype
-
MIM: 123840
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PPIA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6850] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PPIA
- Gene Name
- peptidylprolyl isomerase A
View Full Product Details