Product Details

SNP ID
rs145506438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:5331132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCACTACTACCCCTTTGTCAG[A/G]AAATCAATGTGACTCTCATCTTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNRC18 PubMed Links

Gene Details

Gene
TNRC18
Gene Name
trinucleotide repeat containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080495.2 Intron NP_001073964.2
XM_017012728.1 Intron XP_016868217.1
XM_017012729.1 Intron XP_016868218.1
XM_017012730.1 Intron XP_016868219.1
XM_017012731.1 Intron XP_016868220.1
XM_017012732.1 Intron XP_016868221.1
XM_017012733.1 Intron XP_016868222.1
XM_017012734.1 Intron XP_016868223.1
XM_017012735.1 Intron XP_016868224.1
XM_017012736.1 Intron XP_016868225.1
XM_017012737.1 Intron XP_016868226.1
XM_017012738.1 Intron XP_016868227.1
XM_017012739.1 Intron XP_016868228.1
XM_017012740.1 Intron XP_016868229.1

View Full Product Details