Product Details

SNP ID
rs148326148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1153156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGATACAGAAGTTGCCGTGACA[C/T]TGGGCACATACCATCTGCAGCATCT
Phenotype
MIM: 610699
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927021 PubMed Links

Gene Details

Gene
LOC101927021
Gene Name
uncharacterized LOC101927021
There are no transcripts associated with this gene.

Gene
ZFAND2A
Gene Name
zinc finger AN1-type containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182491.2 652 Silent Mutation CAA,CAG Q117Q NP_872297.2
XM_017012774.1 652 Silent Mutation CAA,CAG Q117Q XP_016868263.1

View Full Product Details