Product Details

SNP ID
rs148894146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:97020894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTGGAGGTCGGAGGGTGGGCAT[G/T]AGGGTGGTGGCTGAGCGAGCGGGAC
Phenotype
MIM: 600028 MIM: 600030
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DLX5 PubMed Links

Gene Details

Gene
DLX5
Gene Name
distal-less homeobox 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005221.5 2031 Missense Mutation AAT,CAT N238H NP_005212.1
XM_005250185.3 2031 Missense Mutation AAT,CAT N110H XP_005250242.1
XM_017011803.1 2031 Missense Mutation AAT,CAT N110H XP_016867292.1
Gene
DLX6
Gene Name
distal-less homeobox 6
There are no transcripts associated with this gene.

Gene
DLX6-AS1
Gene Name
DLX6 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details