Product Details

SNP ID
rs112798378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:28023406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTCCTCAGCCGTGCATTCTCCG[C/T]GACCTCCCTCAGGCTTCTGTGCAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUGGC PubMed Links

Gene Details

Gene
NUGGC
Gene Name
nuclear GTPase, germinal center associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010906.1 1276 Missense Mutation ACG,GCG T768A NP_001010906.1
XM_011544523.2 1276 Intron XP_011542825.1
XM_011544524.2 1276 Intron XP_011542826.1
XM_011544525.1 1276 Missense Mutation ACG,GCG T381A XP_011542827.1
XM_011544526.2 1276 Intron XP_011542828.1
XM_017013403.1 1276 Intron XP_016868892.1

View Full Product Details