Product Details
- SNP ID
-
rs117550044
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:132024457 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGAGCAGGAGCCACGCTACTGAAG[A/G]TGTTAGCCATTTTCTCTGGGCATCT
- Phenotype
-
MIM: 601658
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OC90
PubMed Links
Gene Details
- Gene
- OC90
- Gene Name
- otoconin 90
View Full Product Details