Product Details

SNP ID
rs142769227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144355992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGACAGTGCTTGGTGTGACCCGG[C/G]TGCCCCTGAGGTTAAGAGAGCACAG
Phenotype
MIM: 609076 MIM: 607882
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBXL6 PubMed Links

Gene Details

Gene
FBXL6
Gene Name
F-box and leucine rich repeat protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012162.3 1524 Missense Mutation ACC,AGC T483S NP_036294.2
NM_024555.5 1524 Missense Mutation ACC,AGC T477S NP_078831.4
Gene
SLC52A2
Gene Name
solute carrier family 52 member 2
There are no transcripts associated with this gene.

Gene
TMEM249
Gene Name
transmembrane protein 249
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252402.2 1524 Intron NP_001239331.1
NM_001252404.2 1524 Intron NP_001239333.1
NM_001280561.1 1524 Intron NP_001267490.1

View Full Product Details