Product Details

SNP ID
rs144271550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:33549434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCGGTGAAAGGCGTGTTGGCAC[A/G]GGAGCACGCCTAACTCATCCTTCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF122 PubMed Links

Gene Details

Gene
RNF122
Gene Name
ring finger protein 122
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024787.3 734 Missense Mutation CCG,CTG P110L NP_079063.2
XM_017013846.1 734 Missense Mutation CCG,CTG P86L XP_016869335.1

View Full Product Details