Product Details

SNP ID
rs147113851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144931081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTTACATTCATGGGGCTTCAGC[A/C]CATTATGAATCCTCTGATGCTGAAT
Phenotype
MIM: 601262
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF16 PubMed Links

Gene Details

Gene
ZNF16
Gene Name
zinc finger protein 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029976.2 1892 Missense Mutation GGG,GTG G569V NP_001025147.2
NM_006958.2 1892 Missense Mutation GGG,GTG G569V NP_008889.2
XM_005272341.2 1892 Missense Mutation GGG,GTG G569V XP_005272398.1
XM_005272342.2 1892 Missense Mutation GGG,GTG G429V XP_005272399.1
XM_011517298.1 1892 Missense Mutation GGG,GTG G569V XP_011515600.1

View Full Product Details