Product Details

SNP ID
rs147651118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:98432982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACAAAAATTATTTGCATCTCTTG[A/G]AGGCATTTCAATTACATATGCAGTA
Phenotype
MIM: 602906 MIM: 605030
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNS2 PubMed Links

Gene Details

Gene
KCNS2
Gene Name
potassium voltage-gated channel modifier subfamily S member 2
There are no transcripts associated with this gene.

Gene
STK3
Gene Name
serine/threonine kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256312.1 2291 Intron NP_001243241.1
NM_001256313.1 2291 Intron NP_001243242.1
NM_006281.3 2291 Intron NP_006272.2
XM_011517248.2 2291 Intron XP_011515550.2
XM_011517251.1 2291 UTR 3 XP_011515553.1
XM_011517252.2 2291 Intron XP_011515554.2
XM_011517253.2 2291 UTR 3 XP_011515555.1
XM_011517255.2 2291 UTR 3 XP_011515557.1
XM_011517257.2 2291 Intron XP_011515559.2
XM_011517258.1 2291 UTR 3 XP_011515560.1
XM_017013756.1 2291 Intron XP_016869245.1
XM_017013757.1 2291 Intron XP_016869246.1
XM_017013758.1 2291 Intron XP_016869247.1
XM_017013759.1 2291 UTR 3 XP_016869248.1
XM_017013760.1 2291 UTR 3 XP_016869249.1
XM_017013761.1 2291 Intron XP_016869250.1
XM_017013762.1 2291 Intron XP_016869251.1
XM_017013763.1 2291 UTR 3 XP_016869252.1
XM_017013764.1 2291 UTR 3 XP_016869253.1
XM_017013765.1 2291 Intron XP_016869254.1

View Full Product Details