Product Details

SNP ID
rs137868712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:98200284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAGATGGTCTTGGTGAAGAAGC[A/G]CAGGTACTGGTAGATTTCCAGGCCA
Phenotype
MIM: 602159 MIM: 609871
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CORO2A PubMed Links

Gene Details

Gene
CORO2A
Gene Name
coronin 2A
There are no transcripts associated with this gene.

Gene
TBC1D2
Gene Name
TBC1 domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267571.1 2728 Missense Mutation CGC,TGC R850C NP_001254500.1
NM_001267572.1 2728 Missense Mutation CGC,TGC R390C NP_001254501.1
NM_018421.3 2728 Missense Mutation CGC,TGC R839C NP_060891.3
XM_011518843.1 2728 Missense Mutation CGC,TGC R390C XP_011517145.1
XM_011518844.2 2728 Intron XP_011517146.1
XM_017014899.1 2728 Missense Mutation CGC,TGC R405C XP_016870388.1

View Full Product Details