Product Details

SNP ID
rs138032749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137087054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCTCGGTTCCTCTCAGTCGGAC[C/T]TCCTGACGCCGCCAGTGGGCGGGGC
Phenotype
MIM: 604346
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAN1B1 PubMed Links

Gene Details

Gene
MAN1B1
Gene Name
mannosidase alpha class 1B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016219.4 128 Missense Mutation CTC,TTC L19F NP_057303.2
XM_006716945.3 128 Missense Mutation CTC,TTC L19F XP_006717008.1
XM_017014239.1 128 Intron XP_016869728.1
Gene
MAN1B1-AS1
Gene Name
MAN1B1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
UAP1L1
Gene Name
UDP-N-acetylglucosamine pyrophosphorylase 1 like 1
There are no transcripts associated with this gene.

View Full Product Details