Product Details

SNP ID
rs140478619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122724802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGTCCATGTACTCAGTGATGAA[A/G]GGCCGGGTAGCCACAGTTATGTACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1L4 PubMed Links

Gene Details

Gene
OR1L4
Gene Name
olfactory receptor family 1 subfamily L member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005235.1 813 Silent Mutation AAA,AAG K271K NP_001005235.1

View Full Product Details