Product Details
- SNP ID
-
rs142053044
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:34729341 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTCATGGTGGCTCCTTTTCACTTG[A/C]CAGATAATTACAATAACGATGAAGA
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM205A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12337278] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM205A
- Gene Name
- family with sequence similarity 205 member A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001141917.1 |
198 |
Missense Mutation |
TGG,TGT |
W29C |
NP_001135389.1 |
View Full Product Details