Product Details

SNP ID
rs144515211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:121343918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGCTGCTAGTGGAACTGCTTTA[A/G]AGTGGCCGGTGCGATTCATAGGTGG
Phenotype
MIM: 133090
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STOM PubMed Links

Gene Details

Gene
STOM
Gene Name
stomatin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270526.1 Intron NP_001257455.1
NM_001270527.1 Intron NP_001257456.1
NM_004099.5 Intron NP_004090.4
NM_198194.2 Intron NP_937837.1

View Full Product Details