Product Details

SNP ID
rs147017920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:78239402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTATTGGTTGCTTGTGTTCTGCA[C/T]AGAGGCACTGAAATCTCAGCTGTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP78 PubMed Links

Gene Details

Gene
CEP78
Gene Name
centrosomal protein 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098802.1 Intron NP_001092272.1
NM_032171.1 Intron NP_115547.1
XM_005252263.4 Intron XP_005252320.1
XM_005252265.2 Intron XP_005252322.1
XM_005252266.4 Intron XP_005252323.1
XM_017015192.1 Intron XP_016870681.1
XM_017015193.1 Intron XP_016870682.1
XM_017015194.1 Intron XP_016870683.1
XM_017015195.1 Intron XP_016870684.1
XM_017015196.1 Intron XP_016870685.1

View Full Product Details