Product Details

SNP ID
rs148562883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:14082067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCAAAGAACGTTATCCTCCAAC[A/C]GGGCACAATAAAACCTTCACTAACA
Phenotype
MIM: 600728
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NFIB PubMed Links

Gene Details

Gene
NFIB
Gene Name
nuclear factor I B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190737.1 9583 UTR 3 NP_001177666.1
NM_001190738.1 9583 UTR 3 NP_001177667.1
NM_001282787.1 9583 UTR 3 NP_001269716.1
NM_005596.3 9583 UTR 3 NP_005587.2
XM_005251467.2 9583 UTR 3 XP_005251524.1
XM_005251468.4 9583 UTR 3 XP_005251525.1
XM_005251469.2 9583 UTR 3 XP_005251526.1
XM_005251470.2 9583 UTR 3 XP_005251527.1
XM_005251471.3 9583 UTR 3 XP_005251528.1
XM_006716773.2 9583 UTR 3 XP_006716836.1
XM_006716774.2 9583 UTR 3 XP_006716837.1
XM_006716775.2 9583 UTR 3 XP_006716838.1
XM_017014739.1 9583 UTR 3 XP_016870228.1
XM_017014740.1 9583 UTR 3 XP_016870229.1
XM_017014741.1 9583 UTR 3 XP_016870230.1
XM_017014742.1 9583 UTR 3 XP_016870231.1

View Full Product Details