Product Details

SNP ID
rs149607683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136717688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGTCCTACTCGGAGCGCTGTCGC[A/G]GCCATGTCTGCCAGGTGGTCATTGT
Phenotype
MIM: 614543
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM69B PubMed Links

Gene Details

Gene
FAM69B
Gene Name
family with sequence similarity 69 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152421.3 400 Missense Mutation AGC,GGC S59G NP_689634.2
XM_006716954.3 400 Missense Mutation AGC,GGC S62G XP_006717017.1
XM_011518238.2 400 Missense Mutation AGC,GGC S104G XP_011516540.1
XM_017014273.1 400 UTR 5 XP_016869762.1
Gene
SNHG7
Gene Name
small nucleolar RNA host gene 7
There are no transcripts associated with this gene.

Gene
SNORA17A
Gene Name
small nucleolar RNA, H/ACA box 17A
There are no transcripts associated with this gene.

Gene
SNORA17B
Gene Name
small nucleolar RNA, H/ACA box 17B
There are no transcripts associated with this gene.

View Full Product Details