Product Details

SNP ID
rs150499184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37756182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAAAATGCTATGAAGTTTAAAG[G/T]AGGAAGAGATCACATGGCTATTAAC
Phenotype
MIM: 616919
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FRMPD1 PubMed Links

Gene Details

Gene
FRMPD1
Gene Name
FERM and PDZ domain containing 1
There are no transcripts associated with this gene.

Gene
TRMT10B
Gene Name
tRNA methyltransferase 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286950.1 Intron NP_001273879.1
NM_001286951.1 Intron NP_001273880.1
NM_001286952.1 Intron NP_001273881.1
NM_001286953.1 Intron NP_001273882.1
NM_001286954.1 Intron NP_001273883.1
NM_144964.3 Intron NP_659401.2
XM_005251375.2 Intron XP_005251432.1
XM_005251379.2 Intron XP_005251436.1
XM_011517735.2 Intron XP_011516037.1
XM_011517736.2 Intron XP_011516038.1
XM_011517738.2 Intron XP_011516040.1
XM_011517739.2 Intron XP_011516041.1
XM_011517742.1 Intron XP_011516044.1
XM_011517743.2 Intron XP_011516045.1
XM_017014313.1 Intron XP_016869802.1

View Full Product Details