Product Details

SNP ID
rs150923091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92385726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCGTATCTCTTCAATGCGGTCC[C/T]GGATGTAACTGGTGTCATTAGCCTT
Phenotype
MIM: 611505 MIM: 602383
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPP PubMed Links

Gene Details

Gene
CENPP
Gene Name
centromere protein P
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012267.2 1070 Intron NP_001012267.1
NM_001286969.1 1070 Intron NP_001273898.1
NM_001286971.1 1070 Intron NP_001273900.1
XM_011518685.2 1070 Intron XP_011516987.1
XM_011518689.1 1070 Intron XP_011516991.1
XM_017014715.1 1070 Intron XP_016870204.1
Gene
OGN
Gene Name
osteoglycin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014057.4 1070 Missense Mutation CAG,CGG Q264R NP_054776.1
NM_024416.4 1070 Missense Mutation CAG,CGG Q322R NP_077727.3
NM_033014.3 1070 Missense Mutation CAG,CGG Q264R NP_148935.1

View Full Product Details