Product Details

SNP ID
rs79968027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:1412794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGACTTTGTCGTCTGGCCAGTCA[C/T]GCAGGATCCGGCACAGGACGTACAG
Phenotype
MIM: 300162 MIM: 400011
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASMTL PubMed Links

Gene Details

Gene
ASMTL
Gene Name
acetylserotonin O-methyltransferase-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173473.1 1635 Missense Mutation CAT,CGT H470R NP_001166944.1
NM_001173474.1 1635 Missense Mutation CAT,CGT H512R NP_001166945.1
NM_004192.3 1635 Missense Mutation CAT,CGT H528R NP_004183.2
XM_005274434.3 1635 Missense Mutation CAT,CGT H528R XP_005274491.1
XM_011546185.2 1635 Missense Mutation CAT,CGT H445R XP_011544487.1
XM_017029919.1 1635 Missense Mutation CAT,CGT H419R XP_016885408.1

View Full Product Details