Product Details

SNP ID
rs140125636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:23988635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTACAGCAAATTCAGAGCGTGG[C/T]ACAGACATATCTGCCATCTGCAGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL15 PubMed Links

Gene Details

Gene
KLHL15
Gene Name
kelch like family member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030624.2 1569 Silent Mutation NP_085127.2
XM_006724518.1 1569 Silent Mutation XP_006724581.1

View Full Product Details