Product Details

SNP ID
rs140245947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130384826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGGGGATCGATAACCTCCAGCCA[C/T]GGCTCCTGCATGTTCCATCTTTCGA
Phenotype
MIM: 300513
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR119 PubMed Links

Gene Details

Gene
GPR119
Gene Name
G protein-coupled receptor 119
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178471.2 712 Missense Mutation ATG,GTG M208V NP_848566.1

View Full Product Details