Product Details

SNP ID
rs140674609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49113725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGTTGCACCAAAGCCCGGCTCC[A/G]TGCTCTGTCCCGGCTCAGCAAATGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPKOW PubMed Links

Gene Details

Gene
GPKOW
Gene Name
G-patch domain and KOW motifs
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015698.5 1406 Missense Mutation CGG,TGG R443W NP_056513.2
XM_017029415.1 1406 Missense Mutation CGG,TGG R458W XP_016884904.1

View Full Product Details