Product Details

SNP ID
rs140682311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49117047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTCACTGAGATCCAAGGTGTTC[C/T]TGTCAAACTCCTGCTGGGAGACAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPKOW PubMed Links

Gene Details

Gene
GPKOW
Gene Name
G-patch domain and KOW motifs
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015698.5 975 Missense Mutation AAG,AGG K299R NP_056513.2
XM_017029415.1 975 Missense Mutation AAG,AGG K314R XP_016884904.1

View Full Product Details