Product Details

SNP ID
rs141413370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135965098 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAATGCATCTTGGTTTGAAGAAT[G/T]TGCTGTATCCGAAGGCCGGAAAAGT
Phenotype
MIM: 300231
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MMGT1 PubMed Links

Gene Details

Gene
MMGT1
Gene Name
membrane magnesium transporter 1
There are no transcripts associated with this gene.

Gene
SLC9A6
Gene Name
solute carrier family 9 member A6
There are no transcripts associated with this gene.

View Full Product Details