Product Details

SNP ID
rs142122427
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135257789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGAGGTCTCCGCATGGCAATCC[A/G]AGGAATTCTTAGAGATCTTTTATTA
Phenotype
MIM: 314997
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF75D PubMed Links

Gene Details

Gene
ZNF75D
Gene Name
zinc finger protein 75D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185063.2 Intron NP_001171992.1
NM_007131.4 Intron NP_009062.2
XM_005262469.1 Intron XP_005262526.1
XM_005262471.3 Intron XP_005262528.1
XM_011531391.1 Intron XP_011529693.1
XM_011531395.2 Intron XP_011529697.1
XM_011531396.2 Intron XP_011529698.1
XM_011531397.2 Intron XP_011529699.1
XM_017029818.1 Intron XP_016885307.1
XM_017029819.1 Intron XP_016885308.1
XM_017029820.1 Intron XP_016885309.1

View Full Product Details