Product Details

SNP ID
rs142839385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:131544150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAACTATCCTCAATGGAGAGACA[C/T]GTTTTTCACATTAGTGCTGATAATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR13H1 PubMed Links

Gene Details

Gene
OR13H1
Gene Name
olfactory receptor family 13 subfamily H member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004486.1 77 Missense Mutation ACG,ATG T26M NP_001004486.1

View Full Product Details