Product Details
- SNP ID
-
rs142917667
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:135287679 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCTTTTCTTCCTTGGAAAAGCTC[A/G]ATGCCATTTCTGTACACTGTGTGTA
- Phenotype
-
MIM: 314997
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF75D
PubMed Links
Gene Details
- Gene
- ZNF75D
- Gene Name
- zinc finger protein 75D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001185063.2 |
2830 |
Intron |
|
|
NP_001171992.1 |
NM_007131.4 |
2830 |
Nonsense Mutation |
CGA,TGA |
R331* |
NP_009062.2 |
XM_005262469.1 |
2830 |
Nonsense Mutation |
CGA,TGA |
R331* |
XP_005262526.1 |
XM_005262471.3 |
2830 |
Nonsense Mutation |
CGA,TGA |
R124* |
XP_005262528.1 |
XM_011531391.1 |
2830 |
Nonsense Mutation |
CGA,TGA |
R331* |
XP_011529693.1 |
XM_011531395.2 |
2830 |
Nonsense Mutation |
CGA,TGA |
R181* |
XP_011529697.1 |
XM_011531396.2 |
2830 |
Nonsense Mutation |
CGA,TGA |
R163* |
XP_011529698.1 |
XM_011531397.2 |
2830 |
Nonsense Mutation |
CGA,TGA |
R160* |
XP_011529699.1 |
XM_017029818.1 |
2830 |
Nonsense Mutation |
CGA,TGA |
R185* |
XP_016885307.1 |
XM_017029819.1 |
2830 |
Nonsense Mutation |
CGA,TGA |
R197* |
XP_016885308.1 |
XM_017029820.1 |
2830 |
Nonsense Mutation |
CGA,TGA |
R160* |
XP_016885309.1 |
View Full Product Details