Product Details

SNP ID
rs145351685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53225830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCAAGTAAAGGAAGGAAATCTT[C/T]ATTTCATCCAAGGTTACAGGTAGAG
Phenotype
MIM: 300522 MIM: 314690
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IQSEC2 PubMed Links

Gene Details

Gene
IQSEC2
Gene Name
IQ motif and Sec7 domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111125.2 5860 Intron NP_001104595.1
NM_001243197.1 5860 Intron NP_001230126.1
NM_015075.1 5860 Intron NP_055890.1
XM_006724579.2 5860 Intron XP_006724642.1
XM_006724580.3 5860 Intron XP_006724643.1
XM_006724581.3 5860 Intron XP_006724644.1
XM_006724582.3 5860 UTR 3 XP_006724645.1
XM_006724583.3 5860 Intron XP_006724646.1
XM_006724584.2 5860 Intron XP_006724647.1
XM_011530773.2 5860 Intron XP_011529075.1
XM_011530774.2 5860 Intron XP_011529076.1
XM_011530776.1 5860 Intron XP_011529078.1
XM_011530777.1 5860 Intron XP_011529079.1
XM_017029359.1 5860 Intron XP_016884848.1
XM_017029360.1 5860 Intron XP_016884849.1
Gene
KDM5C
Gene Name
lysine demethylase 5C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146702.1 5860 Intron NP_001140174.1
NM_001282622.1 5860 Intron NP_001269551.1
NM_004187.3 5860 Intron NP_004178.2
XM_005262035.4 5860 Intron XP_005262092.1
XM_011530824.2 5860 Intron XP_011529126.1
XM_011530825.2 5860 Intron XP_011529127.1
XM_011530826.2 5860 Intron XP_011529128.1
XM_011530827.2 5860 Intron XP_011529129.1
XM_011530828.1 5860 Intron XP_011529130.1
XM_011530829.2 5860 Intron XP_011529131.1
XM_011530830.2 5860 Intron XP_011529132.1
XM_011530831.2 5860 Intron XP_011529133.1
XM_017029886.1 5860 Intron XP_016885375.1
XM_017029887.1 5860 Intron XP_016885376.1
XM_017029888.1 5860 Intron XP_016885377.1
XM_017029889.1 5860 Intron XP_016885378.1

View Full Product Details