Product Details

SNP ID
rs145791688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:72582457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTATTTTTTCCACAGCAATGATGC[C/T]TCCGATGCTATGAATTTCAATATCT
Phenotype
MIM: 300269 MIM: 311870
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HDAC8 PubMed Links

Gene Details

Gene
HDAC8
Gene Name
histone deacetylase 8
There are no transcripts associated with this gene.

Gene
PHKA1
Gene Name
phosphorylase kinase regulatory subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122670.1 3405 Missense Mutation AGC,GGC S1134G NP_001116142.1
NM_001172436.1 3405 Missense Mutation AGC,GGC S1075G NP_001165907.1
NM_002637.3 3405 Missense Mutation AGC,GGC S1147G NP_002628.2
XM_006724661.2 3405 Missense Mutation AGC,GGC S1088G XP_006724724.1

View Full Product Details