Product Details

SNP ID
rs145941565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:68049873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAAGCTGTCGGGCCTCTTTGAAA[C/T]GTTCTCACCTCCTTTATGTTCTTGA
Phenotype
MIM: 300127
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OPHN1 PubMed Links

Gene Details

Gene
OPHN1
Gene Name
oligophrenin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002547.2 Intron NP_002538.1
XM_005262270.1 Intron XP_005262327.1
XM_006724653.2 Intron XP_006724716.1
XM_011530961.1 Intron XP_011529263.1
XM_017029555.1 Intron XP_016885044.1

View Full Product Details