Product Details

SNP ID
rs147393092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:16124195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGTTCCAAACCTGTTCATTTCCA[A/G]TCTGGCTTTGGGAGACCTGCTCCTC
Phenotype
MIM: 305670
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRPR PubMed Links

Gene Details

Gene
GRPR
Gene Name
gastrin releasing peptide receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005314.2 895 Missense Mutation AAT,AGT N81S NP_005305.1

View Full Product Details