Product Details

SNP ID
rs148451577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:109654712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCATAGTAGAGCCCGGGCAAGTG[C/T]CCACTAAGCCTGTGCCTCTGAAGAT
Phenotype
MIM: 300157
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSL4 PubMed Links

Gene Details

Gene
ACSL4
Gene Name
acyl-CoA synthetase long-chain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318509.1 Intron NP_001305438.1
NM_001318510.1 Intron NP_001305439.1
NM_004458.2 Intron NP_004449.1
NM_022977.2 Intron NP_075266.1
XM_005262109.2 Intron XP_005262166.1
XM_006724635.2 Intron XP_006724698.1
XM_011530888.2 Intron XP_011529190.1
XM_011530889.2 Intron XP_011529191.1

View Full Product Details