Product Details

SNP ID
rs150104530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:35919897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTCGTCCCCCGGGATATGGATA[G/T]CTCGGGTAGAGACATGCAGCTGCGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CFAP47 PubMed Links

Gene Details

Gene
CFAP47
Gene Name
cilia and flagella associated protein 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304548.1 157 Missense Mutation AGC,ATC S33I NP_001291477.1
NM_152632.3 157 Missense Mutation AGC,ATC S33I NP_689845.2
XM_017029452.1 157 Missense Mutation AGC,ATC S33I XP_016884941.1
XM_017029453.1 157 Missense Mutation AGC,ATC S33I XP_016884942.1

View Full Product Details