Product Details

SNP ID
rs1064181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:28795270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGGACATGTCACTGAATGGGG[G/A]AAGGGAACCCCCGTATCCACAGTCA
Phenotype
MIM: 194355 MIM: 612062
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CCDC117 PubMed Links

Gene Details

Gene
CCDC117
Gene Name
coiled-coil domain containing 117
There are no transcripts associated with this gene.

Gene
XBP1
Gene Name
X-box binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079539.1 Intron NP_001073007.1
NM_005080.3 Intron NP_005071.2
Gene
ZNRF3
Gene Name
zinc and ring finger 3
There are no transcripts associated with this gene.

View Full Product Details