Product Details

SNP ID
rs11538092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38221005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAATCTAGCACTGGACAAAGGT[A/G]GACAGGGGAGGGATGCAGGCGGGAA
Phenotype
MIM: 604877
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAFF PubMed Links

Gene Details

Gene
MAFF
Gene Name
MAF bZIP transcription factor F
There are no transcripts associated with this gene.

Gene
SNORA92
Gene Name
small nucleolar RNA, H/ACA box 92
There are no transcripts associated with this gene.

Gene
TMEM184B
Gene Name
transmembrane protein 184B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195071.1 1897 UTR 3 NP_001182000.1
NM_001195072.1 1897 UTR 3 NP_001182001.1
NM_012264.4 1897 UTR 3 NP_036396.2
XM_011530112.2 1897 UTR 3 XP_011528414.2
XM_011530113.1 1897 UTR 3 XP_011528415.1
XM_011530114.1 1897 UTR 3 XP_011528416.1
XM_011530115.2 1897 UTR 3 XP_011528417.1
XM_011530116.2 1897 Intron XP_011528418.2
XM_011530117.1 1897 UTR 3 XP_011528419.1
XM_011530118.1 1897 UTR 3 XP_011528420.1
XM_011530119.1 1897 UTR 3 XP_011528421.1
XM_017028754.1 1897 UTR 3 XP_016884243.1
XM_017028755.1 1897 UTR 3 XP_016884244.1
XM_017028756.1 1897 UTR 3 XP_016884245.1
XM_017028757.1 1897 UTR 3 XP_016884246.1
XM_017028758.1 1897 Intron XP_016884247.1
XM_017028759.1 1897 UTR 3 XP_016884248.1

View Full Product Details