Product Details

SNP ID
rs11553055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:1403432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTTCAGTGACTGCATCAGGGCG[T/C]GCTGCGCCACCCTCTTCTCCTCATC
Phenotype
MIM: 300162 MIM: 400011
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ASMTL PubMed Links

Gene Details

Gene
ASMTL
Gene Name
acetylserotonin O-methyltransferase-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173473.1 1755 Missense Mutation CAC,CGC H510R NP_001166944.1
NM_001173474.1 1755 Missense Mutation CAC,CGC H552R NP_001166945.1
NM_004192.3 1755 Missense Mutation CAC,CGC H568R NP_004183.2
XM_005274434.3 1755 Missense Mutation CAC,CGC H568R XP_005274491.1
XM_011546185.2 1755 Missense Mutation CAC,CGC H485R XP_011544487.1
XM_017029919.1 1755 Missense Mutation CAC,CGC H459R XP_016885408.1

View Full Product Details