Product Details

SNP ID
rs180819697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2029587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATCGCCCATTCGCCCCAGGTGGT[A/G]CAAAGGATCAAGGCTGTGGAGGGGC
Phenotype
MIM: 607997 MIM: 606553
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPW PubMed Links

Gene Details

Gene
NPW
Gene Name
neuropeptide W
There are no transcripts associated with this gene.

Gene
SLC9A3R2
Gene Name
SLC9A3 regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130012.2 357 Silent Mutation GTA,GTG V73V NP_001123484.1
NM_001252073.1 357 Intron NP_001239002.1
NM_001252075.1 357 Intron NP_001239004.1
NM_001252076.1 357 Intron NP_001239005.1
NM_004785.5 357 Silent Mutation GTA,GTG V73V NP_004776.3
XM_005255702.4 357 Intron XP_005255759.1
XM_006720981.1 357 Intron XP_006721044.1
XM_017023894.1 357 Intron XP_016879383.1
XM_017023895.1 357 Silent Mutation GTA,GTG V17V XP_016879384.1

View Full Product Details