Product Details

SNP ID
rs180931918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33292149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCCCTGTGGCCTTGATCCTGG[C/G]AAAGGAGCCCCCTCCTCCCTCACTC
Phenotype
MIM: 605660 MIM: 602306 MIM: 601962 MIM: 611440
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR6834 PubMed Links

Gene Details

Gene
MIR6834
Gene Name
microRNA 6834
There are no transcripts associated with this gene.

Gene
MIR6873
Gene Name
microRNA 6873
There are no transcripts associated with this gene.

Gene
PFDN6
Gene Name
prefoldin subunit 6
There are no transcripts associated with this gene.

Gene
RGL2
Gene Name
ral guanine nucleotide dissociation stimulator like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243738.1 2650 Missense Mutation CCC,GCC P681A NP_001230667.1
NM_004761.4 2650 Missense Mutation CCC,GCC P763A NP_004752.1
XM_011514781.1 2650 Missense Mutation CCC,GCC P656A XP_011513083.1
XM_011514785.2 2650 Intron XP_011513087.1
Gene
TAPBP
Gene Name
TAP binding protein (tapasin)
There are no transcripts associated with this gene.

Gene
WDR46
Gene Name
WD repeat domain 46
There are no transcripts associated with this gene.

View Full Product Details