Product Details

SNP ID
rs181672079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176189957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCGAGATAGCCAACTGCTTGCAC[C/G]TGAATGACACGCAAGTCAAAATCTG
Phenotype
MIM: 142987
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HAGLR PubMed Links

Gene Details

Gene
HAGLR
Gene Name
HOXD antisense growth-associated long non-coding RNA
There are no transcripts associated with this gene.

Gene
HOXD1
Gene Name
homeobox D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024501.2 1066 Missense Mutation CTG,GTG L268V NP_078777.1
XM_005246507.3 1066 UTR 3 XP_005246564.1
Gene
MIR7704
Gene Name
microRNA 7704
There are no transcripts associated with this gene.

View Full Product Details