Product Details
- SNP ID
-
rs181532198
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:31723280 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGTCTCGCTGTCTCTAAGCCGGA[C/T]CTGATCACCTTTTTGGAGCAAAGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF720
PubMed Links
Gene Details
- Gene
- ZNF720
- Gene Name
- zinc finger protein 720
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130913.1 |
357 |
Silent Mutation |
GAC,GAT |
D51D |
NP_001124385.1 |
View Full Product Details