Product Details

SNP ID
rs182058100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39606521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCCCTTGTCGCTCCTCGGCTCGT[C/T]GTCTTCGCCGTCGCCCCAGCTGGCG
Phenotype
MIM: 601725
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEUROD2 PubMed Links

Gene Details

Gene
NEUROD2
Gene Name
neuronal differentiation 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006160.3 279 Missense Mutation AAC,GAC N27D NP_006151.3

View Full Product Details