Product Details

SNP ID
rs182834999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:4859955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAGGTCTGGATGGCTTATGAGA[A/C]ATGTTGAAGAAACAACTCTGGCGAC
Phenotype
MIM: 607436 MIM: 611491
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PAPOLB PubMed Links

Gene Details

Gene
PAPOLB
Gene Name
poly(A) polymerase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020144.4 2040 Missense Mutation TGT,TTT C619F NP_064529.4
Gene
RADIL
Gene Name
Rap associating with DIL domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018059.4 2040 Intron NP_060529.4

View Full Product Details