Product Details

SNP ID
rs182816928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:68537631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCAGGGCAAGGAGGTGGGGTTCA[A/G]TGAGGACAGGGCTGAGCTGGTCTTC
Phenotype
MIM: 611062 MIM: 188830
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM20A PubMed Links

Gene Details

Gene
FAM20A
Gene Name
family with sequence similarity 20 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243746.1 1760 Missense Mutation ACT,ATT T353I NP_001230675.1
NM_017565.3 1760 Missense Mutation ACT,ATT T491I NP_060035.2
XM_006721959.2 1760 Missense Mutation ACT,ATT T353I XP_006722022.1
XM_011524918.2 1760 Intron XP_011523220.1
XM_017024781.1 1760 Intron XP_016880270.1
Gene
PRKAR1A
Gene Name
protein kinase cAMP-dependent type I regulatory subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276289.1 1760 Intron NP_001263218.1
NM_001276290.1 1760 Intron NP_001263219.1
NM_001278433.1 1760 Intron NP_001265362.1
NM_002734.4 1760 Intron NP_002725.1
NM_212471.2 1760 Intron NP_997636.1
NM_212472.2 1760 Intron NP_997637.1
XM_011524983.2 1760 Intron XP_011523285.1
XM_011524984.2 1760 Intron XP_011523286.1
XM_011524985.2 1760 Intron XP_011523287.1

View Full Product Details