Product Details
- SNP ID
-
rs182814920
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:58271701 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAACTGGGTACCGATGATGCAGG[C/T]GAGGAGTGGGCCCACGCGGCCTTTG
- Phenotype
-
MIM: 150205
MIM: 606989
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LPO
PubMed Links
Gene Details
- Gene
- LPO
- Gene Name
- lactoperoxidase
There are no transcripts associated with this gene.
- Gene
- MPO
- Gene Name
- myeloperoxidase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000250.1 |
2161 |
Missense Mutation |
ACC,GCC |
T662A |
NP_000241.1 |
View Full Product Details